Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8, Mouse, Clone: 15-11C5, Abnova™

Catalog No. 89135932 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-932 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-932 Supplier Abnova Corporation Supplier No. MAB5308
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD8.

Specifications

Antigen CD8
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 15-11C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD8.
Dilution Flow Cytometry (1:10) Immunohistochemistry (Frozen sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.01% sodium azide)
Host Species Mouse
Immunogen Native purified CD8 from WAG/Rij spleen cells.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This monoclonal antibody recognizes rat CD8 antigen (MW 34 KDa and 39 KDa) and is reactive with all common inbred strains.
Target Species Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.