Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8, Mouse, Clone: C8/144B, Abnova™

Catalog No. 89108941 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-941 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-941 Supplier Abnova Corporation Supplier No. MAB10739
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CD8.

The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen acts as a corepressor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The coreceptor functions as either a homodimer composed of two alpha chains, or as a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 alpha chain isoforms. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CD8
Applications Immunohistochemistry
Classification Monoclonal
Clone C8/144B
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CD8.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein base, 0.09% sodium azide)
Gene CD8A
Gene Alias CD8/Leu2/MAL/p32
Gene Symbols CD8A
Host Species Mouse
Immunogen CD8.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925
Content And Storage Store at 4°C is stable for 3 years.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.