Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CD8, Clone: SK1, PE,

Catalog No. MABF1678 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1678 500 Tests
MABF1676 25 Tests
MABF1677 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MABF1678 Supplier MilliporeSigma™ Supplier No. MABF1678
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD8 clone: SK1 in Human samples, and it is validated for Flow Cytometry

The SK1 antibody is specific for the 32-34kDa alpha chain of human CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor for antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The SK1 antibody is widely used as a phenotypic marker for CD8 on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells. It is cross-reactive with CD8 in several non-human species, including Baboon, Chimpanzee, Cynomolgus and Rhesus. If used together with an alternative Anti-Human CD8a clone, RPA-T8, the SK1 antibody will not block binding of RPA-T8 to CD8a.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone SK1
Conjugate PE
Formulation Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. P01732
Gene Symbols CD8A; MAL
Host Species Mouse
Immunogen Immunogen information for this antibody is proprietary information.
Purification Method Purified by size-exclusion chromatography
Quantity 500 Tests
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°C to 8°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.