Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80, Mouse, Clone: MEM-233, Abnova™

Catalog No. 89111630 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-630 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-630 Supplier Abnova Corporation Supplier No. MAB3845
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CD80.

The B-lymphocyte activation antigen B7-1 (formerly referred to as B7) provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 (MIM 186760) and CTLA4 (MIM 123890) ligands of T cells.[supplied by OMIM

Specifications

Antigen CD80
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone MEM-233
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CD80.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CD80
Gene Alias CD28LG/CD28LG1/LAB7
Gene Symbols CD80
Host Species Mouse
Immunogen Recombinant Fc fusion protein corresponding to extracellular domain of human CD80.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 941
Test Specificity This antibody reacts with CD80 (B7-1), a 60 KDa single chain type I glycoproteinof immunoglobulin supergene family, expressed on professional antigen-presenting cells,such as dendritic cells, macrophages or activated B lymphocytes.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.