Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81, Mouse, PE, Clone: M38, Abnova™

Catalog No. 89112903 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-903 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-903 Supplier Abnova Corporation Supplier No. MAB6495
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD81.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins. This protein appears to promote muscle cell fusion and support myotube maintenance. Also it may be involved in signal transduction. This gene is localized in the tumor-suppressor gene region and thus it is a candidate gene for malignancies. [provided by RefSeq

Specifications

Antigen CD81
Applications Flow Cytometry
Classification Monoclonal
Clone M38
Conjugate PE
Description Mouse monoclonal antibody raised against native CD81.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD81
Gene Alias S5.7/TAPA1/TSPAN28
Gene Symbols CD81
Host Species Mouse
Immunogen Native purified CD81 from human T-ALL cell line (MOLT-4).
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 975
Test Specificity This antibody reacts with CD81, a 25 KDa member of the tetraspanin family, expressedon majority of cells.
Target Species Feline, Human, Rabbit
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.