Learn More
CD86, Mouse, Clone: 1B3, Abnova™
Mouse monoclonal antibody raised against partial recombinant CD86.
Supplier: Abnova Corporation MAB10541
Description
This gene encodes a type I membrane protein that is a member of the immunoglobulin superfamily. This protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of this protein with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of this protein with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full-length sequences have not been determined. [provided by RefSeq
Specifications
CD86 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
CD86 | |
CD86 | |
Recombinant protein corresponding to human CD86. | |
RUO | |
942 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Western Blot | |
1B3 | |
Mouse monoclonal antibody raised against partial recombinant CD86. | |
In ascites (0.03% sodium azide) | |
B7-2/B70/CD28LG2/LAB72/MGC34413 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.