Learn More
CD86, Mouse, Clone: BU63, Abnova™
Mouse monoclonal antibody raised against native CD86.
Supplier: Abnova Corporation MAB3846
Description
This gene encodes a type I membrane protein that is a member of the immunoglobulin superfamily. This protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of this protein with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of this protein with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full-length sequences have not been determined. [provided by RefSeq
Specifications
| CD86 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CD86 | |
| CD86 | |
| Native purified CD86 from human ARH-77 cell. | |
| RUO | |
| 942 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
| BU63 | |
| Mouse monoclonal antibody raised against native CD86. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| B7-2/B70/CD28LG2/LAB72/MGC34413 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with CD86 (B7-2), a 70 KDa type I transmembrane glycoprotein of immunoglobulin supergene family, expressed on professional antigen-presenting cells, such as dendritic cells, macrophages or activated B lymphocytes. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.