Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8A/CD8B, Mouse, Clone: ICO-31, Abnova™

Catalog No. 89112245 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-245 Supplier Abnova Corporation Supplier No. MAB5409
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD8A/CD8B.

Specifications

Antigen CD8A/CD8B
Applications Flow Cytometry
Classification Monoclonal
Clone ICO-31
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD8A/CD8B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.5% saccharose, 0.2% gelatin, 0.09% sodium azide)
Gene Alias CD8/Leu2/MAL/p32
Gene Symbols CD8A
Host Species Mouse
Immunogen Native from T cytotoxic/supressor cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925/926
Test Specificity CD8 antigen (human). T cytotoxic, suppressor cells.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.