Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8A/CD8B, Mouse, Clone: RFT-8, Abnova™

Catalog No. 89112324 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-324 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-324 Supplier Abnova Corporation Supplier No. MAB5618
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD8A/CD8B.

Specifications

Antigen CD8A/CD8B
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RFT-8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD8A/CD8B.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Alias CD8/Leu2/MAL/p32
Gene Symbols CD8A
Host Species Mouse
Immunogen Native purified CD8A/CD8B from human thymocytes and E-rosetted lymphocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925/926
Test Specificity human CD8.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.