Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9, Mouse, Clone: ALB 6, Abnova™

Catalog No. 89111719 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-719 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-719 Supplier Abnova Corporation Supplier No. MAB4159
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD9.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It can modulate cell adhesion and migration and also trigger platelet activation and aggregation. In addition, the protein appears to promote muscle cell fusion and support myotube maintenance. [provided by RefSeq

Specifications

Antigen CD9
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ALB 6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD9.
Dilution Flow Cytometry (2ug/5x105 cells/test) Immunohistochemistry (Frozen sections) (1:25-1:50) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS (1mg/mL BSA)
Gene CD9
Gene Alias 5H9/BA2/BTCC-1/DRAP-27/GIG2/MIC3/MRP-1/P24/TSPAN29
Gene Symbols CD9
Host Species Mouse
Immunogen Native purified CD9 from human bone marrow malignant cells from acute non-T, non-B leukemia.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 928
Test Specificity The molecular weight of the recognized antigen is 24KDa. Recognizes 22-43% of peripheral non-T cells (B cells). Recognizes blood platelets and a few granulocytic cells of normal bone marrow. Recognizes the distal convoluted tubule and the distal right tubule of nephrons.
Target Species Human
Content And Storage Store at -20°C. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.