Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90.2/Thy-1.2, Mouse, Clone: 1aG4/C5, Abnova™

Catalog No. 89136213 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-213 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-213 Supplier Abnova Corporation Supplier No. MAB6437
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CD90.2/Thy-1.2.

Specifications

Antigen CD90.2/Thy-1.2
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1aG4/C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CD90.2/Thy-1.2.
Dilution Flow Cytometry (1 ug/mL) Functional Study Immunocytochemistry Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, 300mM NaCl, pH 7.4
Host Species Mouse
Immunogen Native purified from C3H/Di mice thymocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes alloantigen Thy-1.2 of mouse Thy-1 (CD90); it does not cross-react with Thy-1.1 alloantigen.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.