Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90/Thy1 Antibody (5E10), PE/Cy5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143082100TESTS
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry.Specifications
CD90/Thy1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 1:10-1:1000 | |
P04216 | |
THY1 | |
The immunogen for this antibody was CD90. | |
100 Tests | |
Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
7070 | |
Store at 4C. Do not freeze. |
Flow Cytometry | |
50000000000 | |
PE-Cyanine5 | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction