Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99, Mouse, Clone: H0361.1, Abnova™

Catalog No. 89108976 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-976 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-976 Supplier Abnova Corporation Supplier No. MAB10774

Mouse monoclonal antibody raised against CD99.

The protein encoded by this gene is a cell surface glycoprotein involved in leukocyte migration, T-cell adhesion, ganglioside GM1 and transmembrane protein transport, and T-cell death by a caspase-independent pathway. In addition, the encoded protein may have the ability to rearrange the actin cytoskeleton and may also act as an oncosuppressor in osteosarcoma. Cyclophilin A binds to CD99 and may act as a signaling regulator of CD99. This gene is found in the pseudoautosomal region of chromosomes X and Y and escapes X-chromosome inactivation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CD99
Applications Immunohistochemistry
Classification Monoclonal
Clone H0361.1
Conjugate Unconjugated
Description CD99 molecule
Formulation In PBS, pH 7.4 (protein base, 0.09% sodium azide)
Gene CD99
Gene Alias MIC2, MIC2X, MIC2Y
Gene Symbols CD99
Host Species Mouse
Immunogen CD99.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4267
Content And Storage Store at 4°C. Stable for 3 years.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.