Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-CD99, Clone: HO36-1.1, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | CD99 |
---|---|
Clone | HO36-1.1 |
Concentration | 0.2mg/mL |
Dilution | Flow Cytometry 5 - 10 ul/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24469202
![]() |
Novus Biologicals
NBP2446920.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
CD99 Monoclonal antibody specifically detects CD99 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
CD99 | |
0.2mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Immunology | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
4267 | |
Purified E-rosette forming cells from human peripheral blood lymphocytes | |
Primary | |
Store at 4C. |
HO36-1.1 | |
Flow Cytometry 5 - 10 ul/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Supernatant | |
RUO | |
Human, Rat | |
12E7, antigen identified by monoclonal antibodies 12E7, F21 and O13, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
CD99 | |
IgM κ | |
Tissue culture supernatant | |
Recognizes a sialoglycoprotein of 27-32kDa, identified as CD99, or MIC2 gene product, or E2 antigen. MIC2 gene is located in the pseudo-autosomal region of the human X and Y chromosome. MIC2 gene encodes two distinct proteins, which are produced by alternative splicing of the CD99 gene transcript and are identified as bands of 30 and 32kDa (p30/32).Although its function is not fully understood, CD99 is implicated in various cellular processes including homotypic aggregation of T cells, upregulation of T cell receptor and MHS molecules, apoptosis of immature thymocytes and leukocyte diapedesis.CD99 is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary. Most pancreatic islet cells, Sertoli cells of the testis, and some endothelial cells express this antigen. Mature granulocytes express very little or no CD99. MIC2 is strongly expressed on Ewings sarcoma cells and primitive peripheral neuroectodermal tumors. This MAb shows a very simi |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title