Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2, Mouse, Clone: POH-1, Abnova™

Catalog No. 89110710 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-710 Supplier Abnova Corporation Supplier No. MAB1921
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant CDC2.

The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF), which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Mitotic cyclins stably associate with this protein and function as regulatory subunits. The kinase activity of this protein is controlled by cyclin accumulation and destruction through the cell cycle. The phosphorylation and dephosphorylation of this protein also play important regulatory roles in cell cycle control. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CDC2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone POH-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant CDC2.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 500mM NaCl, pH 7.2 (0.01% sodium azide)
Gene CDC2
Gene Alias CDC28A/CDK1/DKFZp686L20222/MGC111195
Gene Symbols CDC2
Host Species Mouse
Immunogen Recombinant protein corresponding to human CDC2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 983
Test Specificity This protein A purified mouse monoclonal antibody reacts specifically with p34 cdc2 in human tissues and cell lines. This antibody is not cross reactive with other cyclin dependent kinases.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.