Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDH1, Mouse, Clone: 36B5, Abnova™
Mouse monoclonal antibody raised against partial recombinant CDH1.
Supplier: Abnova Corporation MAB9706
Specifications
CDH1 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (1:25-1:50) The optimal working dilution should be determined by the end user. | |
Mouse | |
3 x 1 mL | |
Primary | |
Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing. | |
IgG1 |
Immunohistochemistry | |
36B5 | |
Mouse monoclonal antibody raised against partial recombinant CDH1. | |
Lyophilized from tissue culture supernatant (0.09% sodium azide) | |
Recombinant protein corresponding to the N-terminal external region of CDH1. | |
RUO | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction