Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdh1, Mouse, Clone: M168, Abnova™

Catalog No. 89110403 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-403 Supplier Abnova Corporation Supplier No. MAB1388
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Cdh1.

Specifications

Antigen Cdh1
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M168
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Cdh1.
Dilution ELISA (1:2000) Immunocytochemistry (1:250) Immunoprecipitation (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene Cdh1
Gene Alias AA960649/Ecad/L-CAM/MGC107495/UVO/Um
Gene Symbols Cdh1
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of mouse Cdh1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12550
Test Specificity This sequence is highly conserved in human and rat E-cadherin.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.