Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH2, Mouse, Clone: IAR06, Abnova™

Catalog No. 89113688 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-688 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-688 Supplier Abnova Corporation Supplier No. MAB9559

Mouse monoclonal antibody raised against partial recombinant CDH2.

This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry. At certain central nervous system synapses, presynaptic to postsynaptic adhesion is mediated at least in part by this gene product. [provided by RefSeq

Specifications

Antigen CDH2
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone IAR06
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CDH2.
Dilution Immunohistochemistry (1:100) Western Blot (1:250-1:1000) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.09% sodium azide)
Gene CDH2
Gene Alias CD325/CDHN/CDw325/NCAD
Gene Symbols CDH2
Host Species Mouse
Immunogen Recombinant protein corresponding to 160 amino acids of the C-terminu of human CDH2.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1000
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.