Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH3, Mouse, Clone: NCC-CAD-299, Abnova™

Catalog No. 89113446 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-446 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-446 Supplier Abnova Corporation Supplier No. MAB8363

Mouse monoclonal antibody raised against native CDH3.

This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. This gene is located in a six-cadherin cluster in a region on the long arm of chromosome 16 that is involved in loss of heterozygosity events in breast and prostate cancer. In addition, aberrant expression of this protein is observed in cervical adenocarcinomas. Mutations in this gene have been associated with congential hypotrichosis with juvenile macular dystrophy. [provided by RefSeq

Specifications

Antigen CDH3
Applications Immunohistochemistry (Frozen), Inhibition Assays, Western Blot
Classification Monoclonal
Clone NCC-CAD-299
Conjugate Unconjugated
Description cadherin 3, type 1, P-cadherin (placental)
Formulation Lyophilized from 10mM PBS, pH 7.4 (1% BSA)
Gene CDH3
Gene Alias CDHP, HJMD, PCAD
Gene Symbols CDH3
Host Species Mouse
Immunogen Native purified CDH3 from human epidermal carcinoma cell line A431.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1001
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized waterstore at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.