Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDIPT, Mouse, Clone: 1F8, Abnova™

Catalog No. 89106309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-309 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-309 Supplier Abnova Corporation Supplier No. H00010423M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CDIPT.

Phosphatidylinositol breakdown products are ubiquitous second messengers that function downstream of many G protein-coupled receptors and tyrosine kinases regulating cell growth, calcium metabolism, and protein kinase C activity. Two enzymes, CDP-diacylglycerol synthase and phosphatidylinositol synthase, are involved in the biosynthesis of phosphatidylinositol. Phosphatidylinositol synthase, a member of the CDP-alcohol phosphatidyl transferase class-I family, is an integral membrane protein found on the cytoplasmic side of the endoplasmic reticulum and the Golgi apparatus. [provided by RefSeq

Sequence: MPDENIFLFVPNLIGYARIVFAIISFYFMPCCPLTASSFYLLSGLLDAFDGHAARALNQGTRFGAMLDMLTDRCSTMCLLVNLALLYPGATLFFQISMSLDVASHWLHLHSSVVRGSESHKMIDLSGNPVLRIYYTSRPALFTLCAGNELFYCLLYLFHFSEGPLVGSVGLFRMGLWVTAPIALLKSLISVIHLITAARNMAALDAADRAKKK

Specifications

Antigen CDIPT
Applications ELISA
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant CDIPT.
Formulation ascites with no preservative
Gene CDIPT
Gene Accession No. BC001444
Gene Alias MGC1328/PIS/PIS1
Gene Symbols CDIPT
Host Species Mouse
Immunogen CDIPT (AAH01444,1 a.a. ∽ 213 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10423
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.