Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK7, Mouse, Clone: MO-1, Abnova™

Catalog No. 89110670 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-670 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-670 Supplier Abnova Corporation Supplier No. MAB1855
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CDK7.

The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle. [provided by RefSeq

Specifications

Antigen CDK7
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MO-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CDK7.
Dilution Western Blot (1-10 ug/mL) Immunohistochemistry (1-10 ug/mL) Immunoprecipitation (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene CDK7
Gene Alias CAK1/CDKN7/MO15/STK1/p39MO15
Gene Symbols CDK7
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of human CDK7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1022
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.