Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CDKL5, Clone: 8F3.1,

Catalog No. MABS1132 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1132 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1132 Supplier MilliporeSigma™ Supplier No. MABS1132
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CDKL5 clone: 8F3.1 in Mouse, Rat samples, and it is validated for Western Blotting

Cyclin-dependent kinase-like 5 (EC 2.7.11.22; UniProt O76039; also known as Serine/threonine-protein kinase 9) is encoded by the CDKL5 (also known as EIEE2, STK9) gene (Gene ID 6792) in human. CDKL5/STK9 is a serine/threonine protein kinase widely distributed in all tissues, with highest levels in thymus, testes, and brain, where it is found localized in the nucleus and dendrite of neurons. CDKL5/STK9 expression is found peaked in the early postnatal period and Cdkl5-knockdown in mice leads to autistic-like behavior, dendritic hypotrophy and impairment of neural circuit communication. Likewise, human CDKL5 gene mutations are linked to X-linked neurological developmental disorders involving severe mental retardation and seizures appearing in the first postnatal months. Point mutations in CDKL5 have also been reported in patients with atypical variants of Rett syndrome (RTT), infantile spasms and severe neurodevelopmental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKL5
Applications Western Blot
Classification Monoclonal
Clone 8F3.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O76039
Gene Symbols CDKL5; EIEE2; STK9
Host Species Mouse
Immunogen GST-tagged recombinant human CDKL5 internal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001032420
Target Species Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.