Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdkn1b, Mouse, Clone: DCS-72, Abnova™

Catalog No. 89110771 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-771 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-771 Supplier Abnova Corporation Supplier No. MAB2029

Mouse monoclonal antibody raised against full-length recombinant Cdkn1b.

Specifications

Antigen Cdkn1b
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DCS-72
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant Cdkn1b.
Dilution Western Blot (5 to 10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene Cdkn1b
Gene Alias AA408329/AI843786/Kip1/p27/p27Kip1
Gene Symbols Cdkn1b
Host Species Mouse
Immunogen Recombinant protein corresponding to full length mouse Cdkn1b.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12576
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.