Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2, Mouse, Clone: 1C19, Abnova™

Catalog No. 89104270 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-270 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-270 Supplier Abnova Corporation Supplier No. H00001045M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CDX2.

The level and beta-cell specificity of insulin gene expression are regulated by a set of nuclear proteins that bind to specific sequences within the promoter of the insulin gene (INS; MIM 176730) and interact with RNA polymerase to activate or repress transcription. The proteins LMX1 (MIM 600298) and CDX3 are homeodomain proteins that bind an A/T-rich sequence in the insulin promoter and stimulate its transcription (German et al., 1994 [PubMed 7698771]).[supplied by OMIM

Sequence: MYVSYLLDKDVSMYPSSVRHSGGLNLAPQNFVSPPQYPDYGGYHVAAAAAAAANLDSAQSPGPSWPAAYGAPLREDWNGYAPGGAAAAANAVAHGPNGGSPAAAMGYSSPADYHPHHHPHHHPHHPAAAPSCASGLLQTLNPGPPGPAATAAAEQLSPGGQRRNLCEWMRKPAQQSLGSQVKTRTKDKYRVVYTDHQRLELEKEFHYSRYITIRRKAELAATLGLSERQVKIWFQNRRAKERKINKKKLQQQQQQQPPQPPPPPPQPPQPQPGPLRSVPEPLSPVSSLQASVSGSVPGVLGPTGGVLNPTVTQ

Specifications

Antigen CDX2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C19
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant CDX2.
Formulation PBS with no preservative; pH 7.4
Gene CDX2
Gene Accession No. BC014461
Gene Alias CDX-3/CDX3
Gene Symbols CDX2
Host Species Mouse
Immunogen CDX2 (AAH14461.1,1 a.a. ∽ 313 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1045
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.