Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEA, Mouse, Clone: II135F10 C4, Abnova™

Catalog No. 89135499 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-499 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-499 Supplier Abnova Corporation Supplier No. MAB3675
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CEA.

Specifications

Antigen CEA
Applications ELISA
Classification Monoclonal
Clone II135F10 C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CEA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In BBS
Host Species Mouse
Immunogen Native purified human CEA.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for Carcino Embryonic Antigen.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.