Learn More
Specifications
Specifications
| Antigen | CEA |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | PARLAM4 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against native CEA. |
| Dilution | Immunohistochemistry (1:5-1:10) The optimal working dilution should be determined by the end user. |
| Formulation | In Tris-HCl (0.09% sodium azide) |
| Host Species | Mouse |
| Immunogen | Native purified CEA from membrane of human colon carcinoma cells. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.