Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CEACAM1, Clone: 11-1H,

Catalog No. MABT365MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT365MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT365MI Supplier MilliporeSigma™ Supplier No. MABT365
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CEACAM1 clone: 11-1H in Rat samples, and it is validated for ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blotting

The carcinoembryonic antigen- (CEA-) related cell adhesion molecules (CEACAMs) constitute a 12-member subgroup of the CEA family of immunoglobulin-related proteins first described in 1965 (PMID 4953873). CEACAMs are reported to participate in diverse physiological processes, including cell adhesion, differentiation, proliferation, and survival, as well as in carcinogenesis and bacterial pathogenesis. CEACAM molecules from neighbouring cells can interact via their respective extracellular N-terminal IgV-like domain and mediate cell-cell adhesion through trans-oligomerization. CEACAM molecules within the same cell can also undergo transmembrane domain-mediated cis-oligomerization, an event important for sustaining downstream cellular signaling. Athough CEACAM1 is shown to utilize its cytoplasmic domain for transducing cellular signaling, not all CEACAM members are expressed with a significant cytoplasmic domain, and others (CEACAM5/6/7/8) are GPI-anchored without even a transmembrane domain. (PMID 4735478). Rodent Ceacam1 (also known as C-CAM 105, Cell-CAM 105, Ecto-ATPase, GP110, pp120; UniProt P16573) is encoded by the Ceacam1 gene (NCBI Gene ID 81613). As is the case with its human counterpart (P13688), there exisit a long (canonical; 519aa) and a short (458aa) form with a reduced cytoplamic domain as a result of alternate splicing. An additional 10 splice variants are also reported for human CEACAM1. Clone 11-1H does not cross-react with human CEACAM1/3/4/5/6/7/8 or mouse CEACAM1/2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM1
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 11-1H
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P16573
Gene Symbols Ceacam1
Host Species Mouse
Immunogen Recombinant protein corresponding to the extracellular, excluding N domain of rat CEACAM1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001029032
Target Species Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.