Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM3, Mouse, Clone: 2A71C2, Abnova™

Catalog No. 89112237 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-237 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-237 Supplier Abnova Corporation Supplier No. MAB5380
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant CEACAM3.

This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq

Specifications

Antigen CEACAM3
Applications ELISA
Classification Monoclonal
Clone 2A71C2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant CEACAM3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide)
Gene CEACAM3
Gene Alias CD66D/CEA/CGM1/MGC119875/W264/W282
Gene Symbols CEACAM3
Host Species Mouse
Immunogen Recombinant protein corresponding to human CEACAM3.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1084
Test Specificity This antibody reacts with CEA. There is no cross reactivity with human NCA, hCG and PSA.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.