Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5, Mouse, Clone: SPM506, Abnova™

Catalog No. 89110813 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-813 Supplier Abnova Corporation Supplier No. MAB2080

Mouse monoclonal antibody raised against native CEACAM5.

Specifications

Antigen CEACAM5
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SPM506
Concentration Lot specific
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CEACAM5.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:200) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Gene CEACAM5
Gene Alias CD66e/CEA/DKFZp781M2392
Gene Symbols CEACAM5
Host Species Mouse
Immunogen Native purified human CEACAM5.
Purification Method Protein A purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1048
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.