Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5, Mouse, Clone: SPM541, Abnova™

Catalog No. 89110181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-181 Supplier Abnova Corporation Supplier No. MAB12037
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CEACAM5.

Specifications

Antigen CEACAM5
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SPM541
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CEACAM5.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS (0.05% BSA, 0.05% sodium azide)
Gene CEACAM5
Gene Alias CD66e/CEA/DKFZp781M2392
Gene Symbols CEACAM5
Host Species Mouse
Immunogen Human colon carcinoma extract.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1048
Test Specificity This antibody recognizes proteins of 80-200kDa, identified as different members of CEA family.
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.