Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBPB, Mouse, Clone: 47A1, Abnova™

Catalog No. 89108890 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-890 Supplier Abnova Corporation Supplier No. MAB1069
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CEBPB.

The protein encoded by this intronless gene is a bZIP transcription factor which can bind as a homodimer to certain DNA regulatory regions. It can also form heterodimers with the related proteins CEBP-alpha, CEBP-delta, and CEBP-gamma. The encoded protein is important in the regulation of genes involved in immune and inflammatory responses and has been shown to bind to the IL-1 response element in the IL-6 gene, as well as to regulatory regions of several acute-phase and cytokine genes. In addition, the encoded protein can bind the promoter and upstream element and stimulate the expression of the collagen type I gene. [provided by RefSeq

Specifications

Antigen CEBPB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 47A1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CEBPB.
Dilution Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CEBPB
Gene Accession No. NM_005194
Gene Alias C/EBP-beta/CRP2/IL6DBP/LAP/MGC32080/NF-IL6/TCF5
Gene Symbols CEBPB
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-271 of human CEBPB.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1051
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.