Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPJ, Mouse, Clone: 5D5, Abnova™

Catalog No. 89107078 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-078 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-078 Supplier Abnova Corporation Supplier No. H00055835M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CENPJ.

This gene encodes a protein that belongs to the centromere protein family. During cell division, this protein plays a structural role in the maintenance of centrosome integrity and normal spindle morphology, and it is involved in microtubule disassembly at the centrosome. This protein can function as a transcriptional coactivator in the Stat5 signaling pathway, and also as a coactivator of NF-kappaB-mediated transcription, likely via its interaction with the coactivator p300/CREB-binding protein. Mutations in this gene are associated with primary autosomal recessive microcephaly, a disorder characterized by severely reduced brain size and mental retardation. [provided by RefSeq

Sequence: KYTTAARTFPDKKEREEIQTLKQQIADLREDLKRKETKWSSTHSRLRSQIQMLVRENTDLREEIKVMERFRLDAWKRAEAIESSLEVEKKDKLANTSVRFQNSQISSGTQ

Specifications

Antigen CENPJ
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CENPJ.
Formulation ascites with no preservative
Gene CENPJ
Gene Accession No. NM_018451
Gene Alias BM032/CPAP/LAP/LIP1/MCPH6/MGC131581/MGC131582/MGC142222/MGC142224
Gene Symbols CENPJ
Host Species Mouse
Immunogen CENPJ (NP_060921,979 a.a. ∽ 1088 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55835
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.