Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFB, Mouse, Clone: 28A3, Abnova™

Catalog No. 89108128 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-128 Supplier Abnova Corporation Supplier No. MAB0781

Mouse monoclonal antibody raised against native CFB.

This gene encodes complement factor B, a component of the alternative pathway of complement activation. Factor B circulates in the blood as a single chain polypeptide. Upon activation of the alternative pathway, it is cleaved by complement factor D yielding the noncatalytic chain Ba and the catalytic subunit Bb. The active subunit Bb is a serine protease which associates with C3b to form the alternative pathway C3 convertase. Bb is involved in the proliferation of preactivated B lymphocytes, while Ba inhibits their proliferation. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. This cluster includes several genes involved in regulation of the immune reaction. Polymorphisms in this gene are associated with a reduced risk of age-related macular degeneration. The polyadenylation site of this gene is 421bp from the 5' end of the gene for complement component 2. [provided by RefSeq]

Specifications

Antigen CFB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 28A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CFB.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Gene CFB
Gene Alias BF/BFD/CFAB/FB/FBI12/GBG/H2-Bf/PBF2
Gene Symbols CFB
Host Species Mouse
Immunogen Native purified human CFB.
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 629
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.