Learn More
CFL1, Mouse, Clone: 5, Abnova™
Mouse monoclonal antibody raised against full-length recombinant CFL1.
Supplier: Abnova Corporation MAB9908
Description
Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus.[supplied by OMIM
Specifications
CFL1 | |
Monoclonal | |
Unconjugated | |
Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
CFL1 | |
CFL | |
Mouse | |
Affinity purification | |
RUO | |
1072 | |
Human | |
Liquid |
ELISA, Western Blot | |
5 | |
Mouse monoclonal antibody raised against full length recombinant CFL1. | |
In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
NM_005507.2 | |
CFL1 | |
Recombinant protein corresponding to full length human CFL1. | |
100 μg | |
Primary | |
It can expression in BGC823 whole cell lysate. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.