Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFLAR, Mouse, Clone: 5D8, Abnova™

Catalog No. 89110806 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-806 Supplier Abnova Corporation Supplier No. MAB2065
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CFLAR.

Specifications

Antigen CFLAR
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5D8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CFLAR.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CFLAR
Gene Alias CASH/CASP8AP1/CLARP/Casper/FLAME/FLAME-1/FLAME1/FLIP/I-FLICE/MRIT/c-FLIP/c-FLIPL/c-FLIPR/c-FLIPS
Gene Symbols CFLAR
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-376 of human CFLAR.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8837
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.