Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGA, Mouse, Clone: INN-hFSH-132, Abnova™

Catalog No. 89112219 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-219 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-219 Supplier Abnova Corporation Supplier No. MAB5316
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against native CGA.

The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family. [provided by RefSeq

Specifications

Antigen CGA
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone INN-hFSH-132
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CGA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CGA
Gene Alias CG-ALPHA/FSHA/GPHA1/GPHa/HCG/LHA/TSHA
Gene Symbols CGA
Host Species Mouse
Immunogen Native purified CGA from human Follicle Stimulating Hormone (FSH).
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Test Specificity A high affinity antibody recognising the alpha 4 epitope of the alpha-subunit of human Chorionic Gonadotrophin (hCG), hLH, hFSH, hTSH and free alpha-subunit. The recognized epitope comprises amino-acids alpha 15 to alpha 22 of the human sequence. Can be used in combination with INN-hCG-2, 22, 24, 32, 72 and INN-hFSH-6 and 60. This antibody will not cross-react with glycoprotein hormones of various animal species.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.