Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGB, Mouse, Clone: 094-10627, Abnova™

Catalog No. 89111734 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-734 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-734 Supplier Abnova Corporation Supplier No. MAB4214
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CGB.

This gene is a member of the glycoprotein hormone beta chain family and encodes the beta 3 subunit of chorionic gonadotropin (CG). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. CG is produced by the trophoblastic cells of the placenta and stimulates the ovaries to synthesize the steroids that are essential for the maintenance of pregnancy. The beta subunit of CG is encoded by 6 genes which are arranged in tandem and inverted pairs on chromosome 19q13.3 and contiguous with the luteinizing hormone beta subunit gene. [provided by RefSeq

Specifications

Antigen CGB
Applications ELISA
Classification Monoclonal
Clone 094-10627
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CGB.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, pH 7.4 (150mM NaCl, 0.09% sodium azide)
Gene CGB
Gene Alias CGB3/hCGB
Gene Symbols CGB
Host Species Mouse
Immunogen Native purified CGB from human pregnancy urine.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1082
Test Specificity human Chorionic Gonadotropin, beta subunit.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.