Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cgd7_4540, Mouse, Clone: 56g46, Abnova™

Catalog No. 89113643 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-643 Supplier Abnova Corporation Supplier No. MAB8914
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant cgd7_4540.

Specifications

Antigen cgd7_4540
Applications ELISA, Western Blot
Classification Monoclonal
Clone 56g46
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant cgd7_4540.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene cgd7_4540
Gene Symbols cgd7_4540
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 9-156 of cgd7_4540.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3371947
Test Specificity This antibody recognizes recombinant protein CP15.
Target Species C. parvum
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.