Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CH25H, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105994 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-994 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-994 Supplier Abnova Corporation Supplier No. H00009023A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CH25H.

This is an intronless gene that is involved in cholesterol and lipid metabolism. The encoded protein is a membrane protein and contains clusters of histidine residues essential for catalytic activity. Unlike most other sterol hydroxylases, this enzyme is a member of a small family of enzymes that utilize diiron cofactors to catalyze the hydroxylation of hydrophobic substrates. [provided by RefSeq

Sequence: VNIWLSVEDHSGYNFPWSTHRLVPFGWYGGVVHHDLHHSHFNCNFAPYFTHWDKILGTLRTASVPAR

Specifications

Antigen CH25H
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CH25H.
Formulation 50% glycerol
Gene CH25H
Gene Accession No. NM_003956
Gene Alias C25H
Gene Symbols CH25H
Host Species Mouse
Immunogen CH25H (NP_003947, 206 a.a. to 272 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9023
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.