Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104280 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-280 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-280 Supplier Abnova Corporation Supplier No. H00001106A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant CHD2.

The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq

Sequence: MMRNKDKSQEEDSSLHSNASSHSASEEASGSDSGSQSESEQGSDPGSGHGSESNSSSESSESQSESESESAGSKSQPVLPEAKEKPASKKERIADVKKMWEEYPDVYGVRRSNRSRQEPSRFNIKEEASSGSESGSPKRRGQRQLKKQEKWKQEPSEDEQEQGTSAESEPEQKKVKARRPVPRRTVPKPRVKKQPKTQRGKRKKQDSSDEDDDDDEAPKRQTRRRAAKNVSYKEDDDFETDSDDLIEMTGEGVDEQQDNSETIEKVLDSRLGKKGATGASTTVYAIEANGDPSGDFDTEKDEGEIQYLIKWKGWSYIHSTWESEESLQQQKVKGLKKLENFKKKEDEIKQWLGKVSPEDVEYFNCQQELASELNKQYQIVERVIAVKTSKSTLGQTDFPAHSRKPAPSNEPEYLCKWMGLPYSECSWEDEALIGKKFQNCIDSFHSRNNSKTIPTRECKALKQRPRFVALKKQPAYLGGENLELRDYQLEGLNWLAHSWCK

Specifications

Antigen CHD2
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant CHD2.
Formulation 50% glycerol
Gene CHD2
Gene Accession No. BC007347
Gene Alias DKFZp547I1315/DKFZp686E01200/DKFZp781D1727/FLJ38614
Gene Symbols CHD2
Host Species Mouse
Immunogen CHD2 (AAH07347, 1 a.a. to 501 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1106
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.