Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholera toxin (Beta-subunit), Mouse, Clone: 3D11, Abnova™

Catalog No. 89135233 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-233 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-233 Supplier Abnova Corporation Supplier No. MAB2794
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cholera toxin (Beta-subunit).

Specifications

Antigen Cholera toxin (Beta-subunit)
Applications ELISA
Classification Monoclonal
Clone 3D11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cholera toxin (Beta-subunit).
Dilution ELISA (1:1000000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Beta-subunit of cholera toxin.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with cholera toxin. Reacts also with Escherichia coli heat labile enterotoxin.
Target Species Bacteria, E. coli
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.