Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholera toxin, Mouse, Clone: B8, Abnova™

Catalog No. 89134923 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-923 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-923 Supplier Abnova Corporation Supplier No. MAB1290
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against cholera toxin.

Specifications

Antigen Cholera toxin
Applications ELISA
Classification Monoclonal
Clone B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against cholera toxin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.1% gelatin, 0.09% sodium azide)
Host Species Mouse
Immunogen Purified beta-subunit of cholera toxin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with cholera toxin
Content And Storage Store the kit at -20°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.