Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin-4-Sulfate, Mouse, Clone: BE-123, Abnova™

Catalog No. 89136253 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-253 Supplier Abnova Corporation Supplier No. MAB6611
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Chondroitin-4-Sulfate.

Specifications

Antigen Chondroitin-4-Sulfate
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone BE-123
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Chondroitin-4-Sulfate.
Dilution Western Blot (1:1000-1:4000) ELISA (1:400-1:3200) Radioimmunoassay (1:50-1:6400) The optimal working dilution should be determined by the end user.
Formulation In ascites
Host Species Mouse
Immunogen Native purified from mouse proteoglycan.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with both mouse (fetal and adult) and human (fetal and adult) chondroitinase ABC digested cartilage proteoglycans. There is no reaction with native proteoglycans or with proteoglycans digested with either chondroitinase AC or testicular hyaluronidase. Chondroitin-4 sulfate disaccharides successfully inhibit the binding of monoclonal antibody to proteoglycans. Cross-reactivity with mouse, human, bovine, sheep, dog and rat.
Target Species Bovine, Canine, Human, Mouse, Rat, Sheep
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.