Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hCG, Mouse, Clone: 27E8, Abnova™
Mouse monoclonal antibody raised against human chorionic gonadotropin (hCG).
Supplier: Abnova Corporation MAB2833
Specifications
| hCG | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 27E8 | |
| Mouse monoclonal antibody raised against human chorionic gonadotropin (hCG). | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| HCG. | |
| RUO | |
| This antibody specifically recognizes beta-subunit of HCG. No cross-react with LH, TSH, FSH. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction