Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHPF2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106969 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-969 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-969 Supplier Abnova Corporation Supplier No. H00054480B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CHPF2 protein.

Sequence: MRLSSLLALLRPALPLILGLSLGCSLSLLRVSWIQGEGEDPCVEAVGERGGPQNPDSRARLDQSDEDFKPRIVPYYRDPNKPYKKVLRTRYIQTELGSRERLLVAVLTSRATLSTLAVAVNRTVAHHFPRLLYFTGQRGARAPAGMQVVSHGDERPAWLMSETLRHLHTHFGADYDWFFIMQDDTYVQAPRLAALAGHLSINQDLYLGRAEEFIGAGEQARYCHGGFGYLLSRSLLLRLRPHLDGCRGDILSARPDEWLGRCLIDSLGVGCVSQHQGQQYRSFELAKNRDPEKEGSSAFLSAFAVHPVSEGTLMYRLHKRFSALELERAYSEIEQLQAQIRNLTVLTPEGEAGLSWPVGLPAPFTPHSRFEVLGWDYFTEQHTFSCADGAPKCPLQGASRADVGDALETALEQLNRRYQPRLRFQKQRLLNGYRRFDPARGMEYTLDLLLECVTQRGHRRALARRVSLLRPLSRVEILPMPYVTEATRVQLVLPLLVAEAAAAPAFLEAFAANVLEPREHALLTLLLVYGPREGGRGAPDPFLGVKAAAAELERRYPGTRLAWLAVRAEAPSQVRLMDVVSKKHPVDTLFFLTTVWTRPGPEVLNRCRMNAISGWQAFFPVHFQEFNPALSPQRSPPGPPGAGPDPPSPPGADPSRGAPIGGRFDRQASAEGCFYNADYLAARARLAGELAGQEEEEALEGLEVMDVFLRFSGLHLFRAVEPGLVQKFSLRDCSPRLSEELYHRCRLSNLEGLGGRAQLAMALFEQEQANST

Specifications

Antigen CHPF2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CHPF2 protein.
Formulation PBS with no preservative; pH 7.4
Gene CHPF2
Gene Accession No. NM_019015.1
Gene Alias ChSy-3/chPF-2/CSGlcAT/CSGLCA-T
Gene Symbols CHPF2
Host Species Mouse
Immunogen CHPF2 (NP_061888.1,1 a.a. ∽ 772 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54480
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.