Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CHUK, Clone: 3G12H9,

Catalog No. MABE628MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE628MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE628MI Supplier MilliporeSigma™ Supplier No. MABE628
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CHUK clone: 3G12H9 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Inhibitor of nuclear factor kappa-B kinase subunit alpha (CHUK), also known as, IKK-alpha (I-Kappa-B kinase-alpha), is a member of the IKK complex which is composed of IKK-alpha, IKK-beta, IKK-gamma and IKAP. CHUK plays an essential role in the NF-kappa-B signaling pathway which is activated by multiple stimuli such as inflammatory cytokines, bacterial or viral products, DNA damages or other cellular stresses. CHUK plays a key role in the negative feedback of NF-kappa-B canonical signaling to limit inflammatory gene activation. Mutation in CHUK has been associated with Cocoon syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHUK
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3G12H9
Formulation Mouse monoclonal IgG1 ascitic fluid containing 0.03% sodium azide.
Gene Accession No. O15111
Gene Symbols CHUK; IKKA; TCF16
Host Species Mouse
Immunogen Purified recombinant fragment of human CHUK expressed in E. coli.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.