Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIB1, Mouse, Clone: 5A1F5E12, 5A1H7E12, Abnova™

Catalog No. 89113421 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-421 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-421 Supplier Abnova Corporation Supplier No. MAB8297
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CIB1.

The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alpha(IIb)beta(3), which may implicate this protein as a regulatory molecule for alpha(IIb)beta(3). [provided by RefSeq

Specifications

Antigen CIB1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 5A1F5E12, 5A1H7E12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CIB1.
Dilution Western Blot (1:500-1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:2000) Immunohistochemistry (Frozen sections) (1:500-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the end
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene CIB1
Gene Alias CIB/KIP/KIP1/SIP2-28
Gene Symbols CIB1
Host Species Mouse
Immunogen Recombinant protein corresponding to truncated human CIB1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10519
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.