Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CISD1, Mouse, Clone: AT1A8, Abnova™

Catalog No. 89109377 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-377 Supplier Abnova Corporation Supplier No. MAB11208
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CISD1.

CISD1 is a member of the CDGSH domain-containing family and may play a role in the regulation of mitochondrial oxidative capacity (Wiley et al., 2007, 2007 [PubMed 17376863] [PubMed 17584744]).[supplied by OMIM

Specifications

Antigen CISD1
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT1A8
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CISD1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol, 0.02% Sodium Azide.
Gene CISD1
Gene Alias C10orf70/MDS029/MGC14684/ZCD1/mitoNEET
Gene Symbols CISD1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 32-108 of human CISD1.
Purification Method Protein A purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55847
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.