Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CITED4, Mouse, Clone: HT13-2D6.3, Abnova™

Catalog No. 89110951 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-951 Supplier Abnova Corporation Supplier No. MAB2352
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant CITED4.

CITED4 belongs to a family of transcriptional coactivators that bind several proteins, including CREB-binding protein (MIM 600140) and p300 (MIM 602700).[supplied by OMIM

Specifications

Antigen CITED4
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HT13-2D6.3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant CITED4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene CITED4
Gene Symbols CITED4
Host Species Mouse
Immunogen Recombinant fusion protein corresponding to full length human CITED4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163732
Target Species Human
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.