Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrullinated Fibrinogen, Mouse, Clone: 3D1, Abnova™

Catalog No. 89135931 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-931 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-931 Supplier Abnova Corporation Supplier No. MAB5303
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Citrullinated Fibrinogen.

Specifications

Antigen Citrullinated Fibrinogen
Applications ELISA
Classification Monoclonal
Clone 3D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Citrullinated Fibrinogen.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM ammonium bicarbonate buffer
Host Species Mouse
Immunogen A synthetic peptide corresponding to mouse Citrullinated Fibrinogen.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Fibrinogen is a protein produced by the liver which helps stop bleeding by helping blood clots to form. Fibrinogen gets deiminated (conversion from arginin to citrullin) by Peptidyl Arginine Deiminase (PAD) in inflamed joints in patients that develop rheumatoid arthritis. Citrulline, while being an amino acid, is not built into proteins during protein synthesis, as it is not coded for by DNA, yet several proteins are known to contain citrulline. Proteins that normally contain citrulline residues include myelin basic protein (MBP), filaggrin, and several histone proteins, while other proteins, like fibrin and vimentin can get deiminated during cell death and tissue inflammation. Patients with rheumatoid arthritis often (at least 80% of them) develop an immune response against proteins containing citrulline.
Target Species Mouse
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.